
cis-2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)cyclopent-3-ene-1-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1212278-30-2
:
Formula:C₂₁H₁₉NO₄
MDL Number:MFCD11505077
:349.38 g/mol
Melting Pt:127…129 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together