cis-2-tert-Butoxycarbonylamino-cyclooctanecarboxylic acid
Catalog # APOSOR307658-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:271.353 g/mol
Storage Temperature:Ambient
CAS Number:1013980-15-8
Formula:C₁₄H₂₅NO₄
MDL Number:MFCD09475429