Order Entry
Czech Republic
ContactUsLinkComponent
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Catalog # APOSOR307639-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Catalog # APOSOR307639-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :295.374 g/mol
Storage Temperature:Ambient
CAS Number:98661-43-9
Formula:C₁₆H₂₅NO₄
MDL Number:MFCD07366631