Order Entry
Czech Republic
ContactUsLinkComponent
2-(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
2-(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Catalog # APOSOR307638-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Catalog # APOSOR307638-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.269 g/mol
Storage Temperature:Ambient
CAS Number:955287-52-2
Formula:C₁₂H₁₇NO₂
MDL Number:MFCD07366630