2-(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propane-1,3-diol
Catalog # APOSOR307637-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.321 g/mol
Storage Temperature:Ambient
CAS Number:98661-42-8
Formula:C₁₄H₂₁NO₄
MDL Number:MFCD07366629