cis-(2-Methyl-2-methylamino-cyclohexyl)-methanol hydrochloride
Catalog # APOSOR307597-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:193.714 g/mol
Storage Temperature:Ambient
CAS Number:1212377-79-1
Formula:C₉H₁₉NO·ClH
MDL Number:MFCD07783055