Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Phenyl-azetidin-2-one
4-Phenyl-azetidin-2-one
4-Phenyl-azetidin-2-one
APOSOR307589-1G
:
Apollo Scientific
:
4-Phenyl-azetidin-2-one
APOSOR307589-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
147.174 g/mol
:
Ambient
:
5661-55-2
:
C₉H₉NO
:
MFCD02179052