(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-acetic acid
Catalog # APOSOR307542-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.278 g/mol
Storage Temperature:Ambient
CAS Number:68345-67-5
Formula:C₁₃H₁₇NO₄
MDL Number:MFCD00835612