
N-(Pyridin-4-yl)pyridine-4-carboxamide
Catalog # APOSOR307226-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.209 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:64479-78-3
Formula:C₁₁H₉N₃O
MDL Number:MFCD00454168