Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Methyl 1-methyl-1H-indazole-5-carboxylate
Catalog # APOSOR30719-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 1-methyl-1H-indazole-5-carboxylate
Catalog # APOSOR30719-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.199 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1092351-82-0
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD11109401