Order Entry
Czech Republic
ContactUsLinkComponent
2-Oxa-8-azaspiro[4.5]decane hydrochloride 98%
2-Oxa-8-azaspiro[4.5]decane hydrochloride 98%
Catalog # APOSOR306484-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Oxa-8-azaspiro[4.5]decane hydrochloride 98%
Catalog # APOSOR306484-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :177.672 g/mol
Storage Temperature:Ambient
CAS Number:479195-19-2
Formula:C₈H₁₅NO·ClH
MDL Number:MFCD11035930