Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Methoxybenzyl)-1H-pyrazole-4-carboxylic acid 97%
1-(4-Methoxybenzyl)-1H-pyrazole-4-carboxylic acid 97%
Catalog # APOSOR306192-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Methoxybenzyl)-1H-pyrazole-4-carboxylic acid 97%
Catalog # APOSOR306192-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :232.235 g/mol
Storage Temperature:Ambient
CAS Number:1105039-93-7
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD08456928