
1-(1-Benzothiophen-4-yl)piperazine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:913614-18-3
:
Formula:C₁₂H₁₄N₂S·ClH
MDL Number:MFCD26743820
:254.779 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together