Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Catalog # APOSOR305068-250MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Catalog # APOSOR305068-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :344.213 g/mol
Storage Temperature:Ambient
CAS Number:890839-29-9
Formula:C₁₈H₂₅BN₂O₄
MDL Number:MFCD13183032