7-(Hydroxymethyl)-2-azaspiro[4.4]nonan-3-one
Catalog # APOSOR304439-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:169.221 g/mol
Storage Temperature:Ambient
CAS Number:1782480-53-8
Formula:C₉H₁₅NO₂
MDL Number:MFCD28649465