Order Entry
Czech Republic
ContactUsLinkComponent
 
(4R)-6-Azaspiro[2.5]octan-4-ol hydrochloride
Catalog # APOSOR304429-1G
Supplier:  Apollo Scientific
CAS Number:  
(4R)-6-Azaspiro[2.5]octan-4-ol hydrochloride
Catalog # APOSOR304429-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.645 g/mol
Storage Temperature:Ambient
Formula:C₇H₁₃NO·ClH
MDL Number:MFCD23105976