Order Entry
Czech Republic
ContactUsLinkComponent
6-Methoxy-2-azaspiro[3.3]heptane hydrochloride
6-Methoxy-2-azaspiro[3.3]heptane hydrochloride
Catalog # APOSOR304397-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Methoxy-2-azaspiro[3.3]heptane hydrochloride
Catalog # APOSOR304397-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.645 g/mol
Storage Temperature:Ambient
CAS Number:1638761-19-9
Formula:C₇H₁₃NO·ClH
MDL Number:MFCD28501414