Order Entry
Czech Republic
ContactUsLinkComponent
3,3-Dimethylcyclobutan-1-one
3,3-Dimethylcyclobutan-1-one
Catalog # APOSOR304364-1G
Supplier:  Apollo Scientific
CAS Number:  
3,3-Dimethylcyclobutan-1-one
Catalog # APOSOR304364-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :98.143 g/mol
Storage Temperature:Ambient
CAS Number:1192-33-2
Formula:C₆H₁₀O
MDL Number:MFCD22054450