5-Chloro-1H-pyrazolo[3,4-b]pyridine
Catalog # APOSOR303341-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:153.569 g/mol
Storage Temperature:Ambient
CAS Number:1240725-66-9
Formula:C₆H₄ClN₃
MDL Number:MFCD11846318