4,6-Dioxo-2,4,5,6-tetrahydro-1H-benzo[g]imidazo[1,2,3-ij]pteridin-12-ium chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:114853-42-8
:
Formula:C₁₂H₉N₄O₂·Cl
MDL Number:MFCD25948723
:276.678 g/mol
Storage Temperature:Ambient
Frequently Bought Together