5-Bromo-1-methyl-1H-pyrazole-4-carboxylic acid ≥98%
Catalog # APOSOR303319-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.009 g/mol
Storage Temperature:Ambient
CAS Number:54367-67-8
Formula:C₅H₅BrN₂O₂
MDL Number:MFCD22052492