1-Phenyl-5-(pyridin-2-yl)-1,2-dihydropyridin-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:381725-50-4
:
Formula:C₁₆H₁₂N₂O
MDL Number:MFCD21607393
Melting Pt:158 °C
:248.279 g/mol
Storage Temperature:Ambient
Frequently Bought Together