1,3-Diethyl 2-(3,6-dichloro-5-nitropyridin-2-yl)propanedioate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:136888-77-2
:
Formula:C₁₂H₁₂Cl₂N₂O₆
MDL Number:MFCD23136038
:351.139 g/mol
Storage Temperature:Ambient
Frequently Bought Together