3,3-Dimethyl-5-nitroindolin-2-one
Catalog # APOSOR302466-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.198 g/mol
Storage Temperature:Ambient
CAS Number:100511-00-0
Formula:C₁₀H₁₀N₂O₃
MDL Number:MFCD13191795