Ethyl 7-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Catalog # APOSOR302241-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.67 g/mol
Storage Temperature:Ambient
CAS Number:1335053-81-0
Formula:C₁₁H₁₁ClN₂O₂
MDL Number:MFCD25973573