Order Entry
Czech Republic
ContactUsLinkComponent
1,6-Dichloro-2,7-naphthyridine
Irritant
1,6-Dichloro-2,7-naphthyridine
Catalog # APOSOR302178-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,6-Dichloro-2,7-naphthyridine
Catalog # APOSOR302178-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :199.037 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1335053-95-6
Formula:C₈H₄Cl₂N₂
MDL Number:MFCD26403990