Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide 95%
Katalogové číslo APOSOR302149-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.967 g/mol
Storage Temperature:Ambient
CAS Number:1014717-10-2
Formula:C₁₀H₁₃BNO₃·Li
MDL Number:MFCD16038156