
4-Bromo-3-nitro-1H-pyrazole
Catalog # APOSOR302098-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.971 g/mol
Melting Pt:200 °C
:GHS07
Storage Temperature:Ambient
CAS Number:89717-64-6
Formula:C₃H₂BrN₃O₂
MDL Number:MFCD01114992