
4-Chloro-2-methylthieno[3,2-d]pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:319442-16-5
:
Formula:C₇H₅ClN₂S
MDL Number:MFCD09834968
Melting Pt:82…84 °C
:184.646 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together