
Methyl (E)-3-(1H-imidazol-4-yl)prop-2-enoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52363-40-3
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD01823277
:152.151 g/mol
Melting Pt:92…94 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together