Ethyl 2-ethyl-5-methylpyrazole-3-carboxylate 95+%
Katalogové číslo APOSOR300543-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.22 g/mol
Storage Temperature:Ambient
CAS Number:50920-64-4
Formula:C₉H₁₄N₂O₂
MDL Number:MFCD00159650