
4-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1357946-69-0
:
Formula:C₆H₃BrIN₃
MDL Number:MFCD22380433
:323.917 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together