
3,4-Dibromo-1H-pyrazolo[4,3-c]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1357945-30-2
:
Formula:C₆H₃Br₂N₃
MDL Number:MFCD22380430
:276.916 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together