
(1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-5-yl)boronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1562245-02-6
:
Formula:C₁₂H₁₅BN₂O₃
MDL Number:MFCD12922966
:246.07 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together