Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
3-Bromo-4-chloro-1H-pyrazolo[3,4-b]pyridine
Catalog # APOSOR300380-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Bromo-4-chloro-1H-pyrazolo[3,4-b]pyridine
Catalog # APOSOR300380-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.465 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1357945-38-0
Formula:C₆H₃BrClN₃
MDL Number:MFCD22380452