
3-Bromo-5-chloro-1H-pyrazolo[3,4-c]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:245325-30-8
:
Formula:C₆H₃BrClN₃
MDL Number:MFCD13177858
:232.465 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together