
2-Methoxyquinoline-6-carbonitrile
Catalog # APOSOR300253-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:184.194 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:99471-68-8
Formula:C₁₁H₈N₂O
MDL Number:MFCD27921733