
2-Bromo-1-(3-bromo-5-methylphenyl)ethanone
Catalog # APOSOR300245-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
ADR:8,III
:291.967 g/mol
UN:3261
:GHS05
Storage Temperature:Ambient
CAS Number:260430-27-1
Formula:C₉H₈Br₂O
MDL Number:MFCD23711607