
1-(3-Bromo-5-isopropylphenyl)ethanamine hydrochloride
Catalog # APOSOR300231-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:278.616 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:2227272-85-5
Formula:C₁₁H₁₆BrN·ClH
MDL Number:MFCD31561265