Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3-Bromo-5-isopropylphenyl)ethanone
1-(3-Bromo-5-isopropylphenyl)ethanone
1-(3-Bromo-5-isopropylphenyl)ethanone
1-(3-Bromo-5-isopropylphenyl)ethanone
APOSOR300229-250MG
:
Apollo Scientific
:
1-(3-Bromo-5-isopropylphenyl)ethanone
APOSOR300229-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
241.124 g/mol
:
GHS07
:
Ambient
:
1782470-88-5
:
C₁₁H₁₃BrO
:
MFCD31561263