Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-2-(tetrahydro-2H-pyran-4-yl)isoindolin-1-one
Irritant
5-Bromo-2-(tetrahydro-2H-pyran-4-yl)isoindolin-1-one
Catalog # APOSOR300218-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-2-(tetrahydro-2H-pyran-4-yl)isoindolin-1-one
Catalog # APOSOR300218-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :296.16 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1614234-06-8
Formula:C₁₃H₁₄BrNO₂
MDL Number:MFCD31561251