
6-Ethynyl-8-phenylimidazo[1,2-a]pyrazine
Catalog # APOSOR300214-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:219.241 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:2227272-60-6
Formula:C₁₄H₉N₃
MDL Number:MFCD31561247