6-[(4-Chlorophenyl)thio]imidazo[2,1-b][1,3]thiazole-5-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175277-53-9
:
Formula:C₁₂H₇ClN₂OS₂
MDL Number:MFCD00102585
:294.78 g/mol
Storage Temperature:Ambient
Frequently Bought Together