Order Entry
Czech Republic
ContactUsLinkComponent
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Irritant
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Catalog # APOSOR29682-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Catalog # APOSOR29682-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.22 g/mol
Melting Pt:149…153 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:175277-09-5
Formula:C₉H₁₄N₂O₂
MDL Number:MFCD00084917