2-(2,3-Dichlorophenyl)-1,3-thiazole-4-carbohydrazide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175276-87-6
:
Formula:C₁₀H₇Cl₂N₃OS
MDL Number:MFCD00277206
:288.153 g/mol
Storage Temperature:Ambient
Frequently Bought Together