0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
5-Chlorobenzo[b]thiophene-3-carboxaldehyde
5-Chlorobenzo[b]thiophene-3-carboxaldehyde
5-Chlorobenzo[b]thiophene-3-carboxaldehyde
APOSOR29527-100MG
:
Apollo Scientific
:
5-Chlorobenzo[b]thiophene-3-carboxaldehyde
APOSOR29527-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
196.653 g/mol
:
119…121 °C
:
GHS07
:
Ambient
:
16296-68-7
:
C₉H₅ClOS
:
MFCD02677690