Order Entry
Czech Republic
ContactUsLinkComponent
1-(Fur-2-yl)-3-(4-methylphenyl)prop-2-en-1-one
Irritant
1-(Fur-2-yl)-3-(4-methylphenyl)prop-2-en-1-one
Catalog # APOSOR28027-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(Fur-2-yl)-3-(4-methylphenyl)prop-2-en-1-one
Catalog # APOSOR28027-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :212.244 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:14385-62-7
Formula:C₁₄H₁₂O₂
MDL Number:MFCD00276130