
2,2-Dimethyl-5-nitro-2,3-dihydro-1H-benzimidazole
Catalog # APOSOR27769-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:193.203 g/mol
Melting Pt:239…241 °C
:GHS07
Storage Temperature:Ambient
CAS Number:306935-58-0
Formula:C₉H₁₁N₃O₂
MDL Number:MFCD28963764