
Isopropyl 2-bromobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:59247-52-8
:
Formula:C₁₀H₁₁BrO₂
MDL Number:MFCD00027951
:243.097 g/mol
Boiling Pt:92 °C (4 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together