

2-Bromo-1-phenyl-1-ethanone oxime
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14181-72-7
:
Formula:C₈H₈BrNO
MDL Number:MFCD02990473
:214.059 g/mol
Melting Pt:97…100 °C
UN:1759
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together